Skip to Content
Merck
All Photos(1)

Key Documents

D63809

Sigma-Aldrich

1,6-Dichlorohexane

98%

Synonym(s):

Hexamethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)6Cl
CAS Number:
Molecular Weight:
155.07
Beilstein:
773673
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.457 (lit.)

bp

87-90 °C/15 mmHg (lit.)

density

1.068 g/mL at 25 °C (lit.)

SMILES string

ClCCCCCCCl

InChI

1S/C6H12Cl2/c7-5-3-1-2-4-6-8/h1-6H2

InChI key

OVISMSJCKCDOPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

165.0 - 170.6 °F - closed cup

Flash Point(C)

73.9 - 77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Heyan Jiang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(34), 8280-8285 (2017-03-24)
The effect of chain structure on flexibility and stability of macromolecules containing weak P-Fe metal coordination bonds is studied. Migration insertion polymerization (MIP) of FpC
Mohsen Ghafari et al.
Journal of hazardous materials, 351, 117-123 (2018-03-12)
A novel one-step hyper-cross-linking method, using 1,2-dichloroethane (DCE) and 1,6-dichlorohexane (DCH) cross-linkers, expands the micropore volume of commercial styrenic polymers. Performance of virgin and modified polymers was evaluated by measuring hexane, toluene, and methyl-ethyl-ketone (MEK) adsorption capacity, adsorption/desorption kinetics, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service