D1602
1-Decanethiol
96%
Synonym(s):
DT, Decyl mercaptan, Mercaptan C10
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)9SH
CAS Number:
Molecular Weight:
174.35
Beilstein:
1735223
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
96%
form
liquid
refractive index
n20/D 1.457 (lit.)
n20/D 1.458 (lit.)
bp
114 °C/13 mmHg (lit.)
mp
−26 °C (lit.)
density
0.824 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CCCCCCCCCCS
InChI
1S/C10H22S/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3
InChI key
VTXVGVNLYGSIAR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
1-Decanethiol is an alkanethiol that forms a self-assembled monolayer(SAM) and it can be coated as a protective coating on a variety of substrates.
Application
1-Decanethiol can be blended with mercaptohexanol to modify the surface of silver for surface enhancement Raman scattering (SERS) based applications. It can act as a corrosion inhibitor on copper-nickel (CuNi) alloy with an inhibition efficiency of 91%. Zinc doped copper indium sulfide (ZnCuInS2) can be functionalized with 1-decanethiol as a ligand to improve the charge mobility and luminance of organic light emitting diodes (OLEDs). 1-Decanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for field-effect transistors (FETs).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
208.4 °F - closed cup
Flash Point(C)
98 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Thiol self-assemble layer as inhibitor to protect B10 from seawater corrosion.
Xu F, et al.
Progress in Organic Coatings, 97(1), 82-90 (2016)
Quantum Dot Light-Emitting Diode with Ligand-Exchanged ZnCuInS2 Quantum Dot.
Fukuda T, et al.
Transactions of the Institute of Electronics, Information and Communication Engineers, 100(11), 943-948 (2017)
Extinction coefficient of gold nanoparticles with different sizes and different capping ligands.
Liu X, et al.
Colloids and Surfaces. B, Biointerfaces, 58(1), 3-7 (2007)
Transparent polymer-based SERS substrates templated by a soda can.
Creedon NC, et al.
Sensors and Actuators B, Chemical, 259(1), 64-74 (2018)
In situ STM imaging of individual molecules in two-component self-assembled monolayers of 3-mercaptopropionic acid and 1-decanethiol on Au (111).
Sawaguchi T, et al.
Journal of Electroanalytical Chemistry, 496(1-2), 50-60 (2001)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service