Skip to Content
Merck
All Photos(1)

Key Documents

705233

Sigma-Aldrich

1-Decanethiol

99%

Synonym(s):

DT, Decyl mercaptan, Mercaptan C10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9SH
CAS Number:
Molecular Weight:
174.35
Beilstein:
1735223
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.457 (lit.)
n20/D 1.458 (lit.)

bp

114 °C/13 mmHg (lit.)

mp

−26 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCS

InChI

1S/C10H22S/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3

InChI key

VTXVGVNLYGSIAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arash Samadi et al.
Thyroid : official journal of the American Thyroid Association, 25(5), 494-502 (2015-03-18)
Thyroid hormone (TH) is essential for the developing brain, and because the fetal thyroid develops relatively late in gestation, the maternal TH supply is critical for fetal brain development. However, if the mother has hypothyroidism during pregnancy, fetal brain and
Stefanie Kehrer et al.
Neuropsychologia, 74, 30-36 (2014-12-03)
The posterior parietal cortex (PPC) and the dorsolateral prefrontal cortex (DLPFC) are known to be part of a cortical network involved in visual spatial attention. Top-down control can modulate processing at target and distractor positions over a sequence of trials
Maithe Arruda-Carvalho et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(47), 15793-15803 (2014-11-21)
Proliferation of neural progenitor cells in the subventricular zone leads to the continuous generation of new olfactory granule cells (OGCs) throughout life. These cells synaptically integrate into olfactory bulb circuits after ∼2 weeks and transiently exhibit heightened plasticity and responses
Sofia N Nyström et al.
European journal of immunology, 44(12), 3621-3631 (2014-09-19)
Depletion of Foxp3(+) CD4(+) regulatory T cells (Treg) in adults results in chronic inflammation and autoimmune disease. However, the impact of transient Treg-cell depletion on self-reactive responses is poorly defined. Here, we studied the effect of transient depletion of Treg
Ignacio Melero et al.
Cancer research, 75(3), 497-507 (2014-12-21)
Host responses are increasingly considered important for the efficacious response to experimental cancer therapies that employ viral vectors, but little is known about the specific nature of host responses required. In this study, we investigated the role of host type

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service