Skip to Content
Merck
All Photos(2)

Key Documents

O1858

Sigma-Aldrich

1-Octadecanethiol

98%

Synonym(s):

Mercaptan C18, Octadecyl mercaptan, Stearyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17SH
CAS Number:
Molecular Weight:
286.56
Beilstein:
1811934
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

bp

204-210 °C/11 mmHg (lit.)

mp

30-33 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCS

InChI

1S/C18H38S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3

InChI key

QJAOYSPHSNGHNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Octadecanethiol (ODT) is an alkanethiol with a long carbon chain mainly used as a self-assembled monolayer (SAM) for surface modification of a wide range of materials. It facilitates superhydrophobicity with high wettability and low surface energy.

Application

ODT is used in the surface modification of gold nanoparticle (AuNPs)/Nickel (Ni) foam conjugate for the preparation of a 3D surface enhanced Raman spectroscopy(SERS) substrate for the detection of organic pollutants. It may also be used enhance the superhydrophobic properties for different materials like nano-sized copper films, graphene and silica for different applications like mercury detection and electrochemical analysis.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical investigation of dynamic interfacial processes at 1-octadecanethiol-modified copper electrodes in halide-containing solutions.
Ma HY, et al.
Electrochimica Acta, 48(28), 4277-4289 (2003)
Three-dimensional superhydrophobic surface-enhanced Raman spectroscopy substrate for sensitive detection of pollutants in real environments.
Zhao H, et al.
Journal of Material Chemistry A, 3(8), 4330-4337 (2015)
Fabrication of super-hydrophobic nano-sized copper films by electroless plating.
Liu X, et al.
Thin Solid Films, 518(14), 3731-3734 (2010)
Adhesion and friction studies of microsphere-patterned surfaces in contact with atomic force microscopy colloidal probe.
Zhang X, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 401(28), 90-96 (2012)
Khalid S Salaita et al.
Nano letters, 6(11), 2493-2498 (2006-11-09)
Experiments that utilize structures generated by dip-pen nanolithography (DPN) as positive resists for fabricating nanohole arrays and lithographic masters are described. The technique takes advantage of the difference in desorption potentials for patterned structures made from 16-mercaptohexadecanoic acid (MHA) and

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service