Skip to Content
Merck
All Photos(3)

Key Documents

C80857

Sigma-Aldrich

trans-Cinnamic acid

≥99%

Synonym(s):

trans-3-Phenylacrylic acid, Cinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOH
CAS Number:
Molecular Weight:
148.16
Beilstein:
1905952
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

crystals

bp

300 °C (lit.)

mp

132-135 °C (lit.)

SMILES string

OC(=O)\C=C\c1ccccc1

InChI

1S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H,10,11)/b7-6+

InChI key

WBYWAXJHAXSJNI-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-Cinnamic acid can be used in the synthesis of:
  • A trans-cinnamic acid hydrazide derivative with potent anti-mycobacterial activity.
  • Cinnamate glycerides via homogeneous esterification reaction.
  • Styrene via biocatalytic decarboxylation by plant cell cultures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homogeneous catalytic esterification of glycerol with cinnamic and methoxycinnamic acids to cinnamate glycerides in solventless medium: Kinetic modeling.
Molinero L, et al.
Chemical Engineering Journal, 247(2), 174-182 (2014)
Synthesis and antimycobacterial evaluation of new trans-cinnamic acid hydrazide derivatives.
Carvalho SA, et al.
Bioorganic & Medicinal Chemistry, 18(2), 538-541 (2008)
M Takemoto et al.
Chemical & pharmaceutical bulletin, 49(5), 639-641 (2001-06-01)
A novel method for producing styrenes from trans-cinnamic acids was developed. When trans-cinnamic acid was incubated with plant cell cultures at room temperature, styrene was obtained. 4-Hydroxy-3-methoxystyrene (2a), 3-nitrostyrene (2f) and furan (2g) were synthesized quantitatively.
Marie S Prevost et al.
Journal of medicinal chemistry, 56(11), 4619-4630 (2013-05-21)
Pentameric ligand gated ion channels (pLGICs) mediate signal transduction. The binding of an extracellular ligand is coupled to the transmembrane channel opening. So far, all known agonists bind at the interface between subunits in a topologically conserved "orthosteric site" whose
Stanislava Gorjanović et al.
Journal of agricultural and food chemistry, 60(38), 9573-9580 (2012-09-07)
Hydrogen peroxide scavenging (HPS) activity of unfermented (green, yellow, and white), partially fermented (oolong), and completely fermented (black) tea ( Camellia sinensis ), maté ( Ilex paraguariensis ), and various herbal infusions, as well as individual compounds (flavan-3-ols, flavonols, cinnamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service