173282
Methyl trans-cinnamate
99%
Synonym(s):
trans-Cinnamic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
Assay
99%
form
solid
bp
260-262 °C (lit.)
mp
34-38 °C (lit.)
SMILES string
COC(=O)\C=C\c1ccccc1
InChI
1S/C10H10O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-8H,1H3/b8-7+
InChI key
CCRCUPLGCSFEDV-BQYQJAHWSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Methyl trans-cinnamate has antimicrobial ability.
Application
Methyl trans-cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of chromatography. A, 1487, 14-21 (2017-01-29)
Specific blends of essential oils (BEOs) are promising substitutes for antibiotics to promote livestock performance and to reduce the incidence of intestinal disorders. Microencapsulation of BEOs has shown to improve their stability, bioavailability and to control their release rate once
Journal of agricultural and food chemistry, 52(10), 3065-3068 (2004-05-13)
For the authenticity assessment of (E)-methyl cinnamate from different origins, combustion/pyrolysis-isotope ratio mass spectrometry (C/P-IRMS) was used by an elemental analyzer (EA) and on-line capillary gas chromatography coupling (HRGC-C/P-IRMS). For that reason, (E)-methyl cinnamate self-prepared from synthetic, natural, and semisynthetic
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S113-S119 (2007-11-27)
A toxicologic and dermatologic review of methyl cinnamate when used as a fragrance ingredient is presented.
Archives of toxicology, 49(3-4), 321-330 (1982-03-01)
The urinary mercapturic acid excretion by female rats of methyl atropate (alpha-phenyl methyl acrylate) and methyl cinnamate (beta-phenyl methyl acrylate) has been studied. On the basis of the structures of these mercapturic acids the conclusion can be drawn that these
International journal of food microbiology, 153(1-2), 183-191 (2011-12-06)
The investigation deals with antifungal, antiaflatoxin and antioxidant efficacy of Zanthoxylum alatum Roxb. essential oil (EO), its two major constituents and their comparison with five commonly used organic acid preservatives. The chemical profile of EO, characterized through GC and GC-MS
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service