Skip to Content
Merck
All Photos(2)

Key Documents

139939

Sigma-Aldrich

trans-Stilbene

96%

Synonym(s):

trans-1,2-Diphenylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHC6H5
CAS Number:
Molecular Weight:
180.25
Beilstein:
1616740
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

96%

form

solid

bp

305-307 °C/744 mmHg (lit.)

mp

123-125 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

SMILES string

c1ccc(cc1)\C=C\c2ccccc2

InChI

1S/C14H12/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-12H/b12-11+

InChI key

PJANXHGTPQOBST-VAWYXSNFSA-N

Gene Information

human ... ESR1(2099)
rat ... Esr1(24890)

Looking for similar products? Visit Product Comparison Guide

Application

Commonly employed in transition metal catalyzed asymmetric epoxidation and dihydroxylation.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron Letters, 32, 5761-5761 (1991)
Tetrahedron Letters, 33, 4021-4021 (1992)
The effect of material purity on the optical and scintillation properties of solution-grown trans-stilbene crystals
Carman, Leslie, et al.
Journal of Crystal Growth, 368, 56-61 (2013)
Miłosz Regulski et al.
Bioorganic & medicinal chemistry, 26(1), 141-151 (2017-12-02)
25 new trans-stilbene and trans-stilbazole derivatives were investigated using in vitro and in silico techniques. The selectivity and potency of the compounds were assessed using commercial ELISA test. The obtained results were incorporated into 2D QSAR assay. The most promising
Conformational information from vibrational spectra of styrene, trans-stilbene, and cis-stilbene.
Choi, Cheol Ho, and Miklos Kertesz
The Journal of Physical Chemistry A, 101(20), 3823-3831 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service