Skip to Content
Merck
All Photos(3)

Key Documents

C77007

Sigma-Aldrich

Cholesteryl chloroformate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H45ClO2
CAS Number:
Molecular Weight:
449.11
Beilstein:
3223154
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

liquid crystal

optical activity

[α]27/D −28°, c = 2 in chloroform

mp

115-117 °C (lit.)

transition temp

crystalline phase to cholesteric phase 117.8 °C
cholesteric phase to isotropic phase 125.3 °C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(Cl)=O

InChI

1S/C28H45ClO2/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(31-26(29)30)13-15-27(20,4)25(22)14-16-28(23,24)5/h9,18-19,21-25H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,27+,28-/m1/s1

InChI key

QNEPTKZEXBPDLF-JDTILAPWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinqin Yin et al.
Scientific reports, 6, 38582-38582 (2016-12-08)
N-methyl bromide of lidocaine (QX-314) is a potential local anaesthetic with compromised penetration through cell membranes due to its obligated positive charge. Liposomes have been widely used for drug delivery with promising efficacy and safety. Therefore we investigated the local
Mengliang Wu et al.
Experimental eye research, 197, 108124-108124 (2020-07-01)
Pterygium is a degenerative disease that characterized by excessive fibrovascular proliferation. To reduce the recurrence rate, surgery is the main strategy, in combination with adjacent procedures or adjunctive therapy. One of the most common adjunctive agents, mitomycin C (MMC), is
Xuelei Pang et al.
Organic & biomolecular chemistry, 14(47), 11176-11182 (2016-11-12)
The facile tuning of the fluorescent properties of organogels is highly desirable for optical switches, light-emitting diodes, chemosensors and bioprobes. The design of organic molecules with multiple emission colors but only one molecular platform remains challenging. Herein, a new cholesterol-based
Mengjie Rui et al.
International journal of nanomedicine, 7, 3751-3768 (2012-08-14)
Natural high-density lipoproteins (HDL) possess important physiological functions to the transport of cholesterol from the peripheral tissues to the liver for metabolic degradation and excretion in the bile. In this work, we took advantage of this pathway and prepared two
Elham Pishavar et al.
Biotechnology progress, 36(3), e2952-e2952 (2019-12-18)
IL-12 is a pleiotropic cytokine, which shows an ideal applicant for tumor immunotherapy, because of its features of creating an interconnection between innate (NK cells) and adaptive (cytotoxic T lymphocyte) immunity. IL-12 gene therapy is a useful technique to deliver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service