Skip to Content
Merck
All Photos(2)

Key Documents

128376

Sigma-Aldrich

4-Fluorobenzaldehyde

98%

Synonym(s):

p-Fluorobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CHO
CAS Number:
Molecular Weight:
124.11
Beilstein:
385857
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.521 (lit.)

bp

181 °C/758 mmHg (lit.)

mp

−10 °C (lit.)

density

1.157 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(F)cc1

InChI

1S/C7H5FO/c8-7-3-1-6(5-9)2-4-7/h1-5H

InChI key

UOQXIWFBQSVDPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Fluorobenzaldehyde was used in the preparation of pyrazolopyridine UR-13756.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark C Bagley et al.
Future medicinal chemistry, 2(2), 193-201 (2011-03-24)
UR-13756 is a potent and selective p38 mitogen-activated protein kinase (MAPK) inhibitor, reported to have good bioavailability and pharmacokinetic properties and, thus, is of potential use in the treatment of accelerated aging in Werner syndrome. Irradiation of 2-chloroacrylonitrile and methylhydrazine
Abdul Hafeez et al.
Polymers, 11(9) (2019-09-22)
Bis-aldehyde monomers 4-(4'-formyl-phenoxy)benzaldehyde (3a), 3-methoxy-4-(4'-formyl-phenoxy)benzaldehyde (3b), and 3-ethoxy-4-(4'-formyl-phenoxy)benzaldehyde (3c) were synthesized by etherification of 4-fluorobenzaldehyde (1) with 4-hydroxybenzaldehyde (2a), 3-methoxy-4-hydroxybenzaldehyde (2b), and 3-ethoxy-4-hydroxybenzaldehyde (2c), respectively. Each monomer was polymerized with p-phenylenediamine and 4,4'-diaminodiphenyl ether to yield six poly(azomethine)s. Single crystal
Xuemei Tian et al.
Enzyme and microbial technology, 84, 32-40 (2016-02-02)
The first Novozym 435 lipase-catalyzed Morita-Baylis-Hillman (MBH) reaction with amides as co-catalyst was realized. Results showed that neither Novozym 435 nor amide can independently catalyze the reaction. This co-catalytic system that used a catalytic amount of Novozym 435 with a
Bin-Yu Wen et al.
Journal of Asian natural products research, 21(7), 702-715 (2019-01-01)
Desmosdumotin C (Des C), a natural product isolated from the roots of Desmos dumosus, has shown good antitumor activity. A three dimensional quantitative structure-activity relationship (QSAR) study using the comparative molecular field analysis (CoMFA) method was performed on 32 Des
Stephan Mokesch et al.
Frontiers in chemistry, 8, 209-209 (2020-04-23)
A series of 2-phenylbenzothiazole derivatives and their corresponding organometallic ruthenium(II) and osmium(II) complexes were synthesized, designed to exploit both, the attributes of the half-sandwich transition metal scaffold and the bioactivity spectrum of the applied 2-phenylbenzothiazoles. All synthesized compounds were characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service