Key Documents
BR782150
BRAND® lids for microplates BRAND®plates
plate size 96 wells, with condensation rings
Synonym(s):
Accessories, BRANDplates®, Lids, Microplates
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
Pricing and availability is not currently available.
material
polystyrene
packaging
pack of 100 ea
manufacturer/tradename
BRAND 782150
plate size
96 wells
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
BR782153BR782152BR782151
packaging
pack of 100 ea
packaging
pack of 50 ea
packaging
pack of 50 ea
packaging
pack of 100 ea
manufacturer/tradename
BRAND 782150
manufacturer/tradename
BRAND 782153
manufacturer/tradename
BRAND 782152
manufacturer/tradename
BRAND 782151
plate size
96 wells
plate size
1536 wells
plate size
-
plate size
96 wells
General description
To protect samples, reaction batches or cell cultures from both contamination and evaporation, compatible lids are available for all BRANDplates® microplates. The lids are made of crystal-clear polystyrene. They are optimally matched to the corresponding BRANDplates® microplates, so that plates with lids can also be stacked. The cut-corner sets the orientation of the lid. For 96-well standard plates, lids with condensation rings are available.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)