CLS3935
Corning® multiwell plates, plate lids and sealing mats
universal multiwell plate lid w/ corner notch, polystyrene, black, sterile, 50/cs
Synonym(s):
universal microplate lid, universal microtiter lid, universal multiwell plate lid, universal plate lid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
Recommended Products
material
polystyrene
sterility
sterile
feature
lid
packaging
pack of 25
case of 50
manufacturer/tradename
Corning 3935
size
96 or 384 wells
color
black
Looking for similar products? Visit Product Comparison Guide
General description
- Suitable for fluorescent and other light-sensitive assays
- Suitable for 96 well, 96 half area & 384 well plates
- Lids are made of rigid polystyrene
- Lids have a corner notch on the A1 corner to correspond to the corner notches found on all Corning® microplates
- Lids do not fit Corning 2μL 1536 well microplates
- Sterilized by gamma radiation
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service