CLS3930
Corning® multiwell plates, plate lids and sealing mats
96 well plate lid, rigid polystyrene low evaporation lid w/ corner notch, condensation rings, sterile, 100/cs
Synonym(s):
96 multiwell plate lid, 96 well microplate lid, 96 well microtiter plate lid, 96 well plate lid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
Recommended Products
material
polystyrene
sterility
sterile
feature
lid
packaging
pack of 1 (individually wrapped)
case of 100
manufacturer/tradename
Corning 3930
size
96 wells
color
clear
Looking for similar products? Visit Product Comparison Guide
General description
- Lids are made of rigid polystyrene
- Lids have a corner notch on the A1 corner to correspond to the corner notches found on all Corning® microplates
- Lids do not fit Corning 2 μL 1536 well microplates or 2 mL 96 well storage blocks
- Sterilized by gamma radiation
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service