BR27511
BRAND® USP BLAUBRAND® class AS graduated pipette
capacity 5 mL
Synonym(s):
BLAUBRAND®, Graduated Pipettes, Pipettes, USP
About This Item
Recommended Products
material
AR-Glass
feature
class AS
graduation
type 2 (nominal volume above)
packaging
pack of 12
manufacturer/tradename
BRAND 27511
measuring range
accuracy: 0.05 mL
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Accurate volumetric measurement instrument for precision analyses.
- BRAND® USP BLAUNRAND® class AS graduated pipette of capacity 5 mL, type 2, made of AR-Glas®, is manufactured according to DIN EN ISO 835.
- Calibrated to deliver (TD, Ex).
- Reduced waiting time (5 sec) and simplified handling.
- High volume precision.
- The meniscus needs to be set -only once- directly to the partial volume.
- Optimized flow geometry in the tip ensures reliable liquid dispensing from bulb and graduated pipettes.
- The mechanical resilience of the tip and its fire-polished edges ensure a long service life.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service