Key Documents
BR143260
BRAND® special laboratory tubing
I.D. × O.D. × W 4 mm × 6 mm × 1 mm, L 20 m
Synonym(s):
Special laboratory tubing, Tubing
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
material
PVC
manufacturer/tradename
BRAND 143260
I.D. × O.D. × W
4 mm × 6 mm × 1 mm
L
20 m
Related Categories
General description
Particularly high-grade material, manufactured to a first-class standard. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies. Autoclavable (121 °C).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)