BR143362
BRAND® tubing
I.D. 8, O.D. 12, W 2.0 mm
Synonym(s):
Tubing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
silicone
manufacturer/tradename
BRAND 143362
I.D.
8
L
25 m
O.D.
12
W
2.0 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The BRAND® tubing is translucent, allowing for easy monitoring of fluid flow. It is autoclavable at 121 °C, making it suitable for sterilization in laboratory settings. However, it is important to note that the tubing has limited chemical resistance when used with concentrated acids, so caution should be exercised in such applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service