Skip to Content
Merck
All Photos(3)

Key Documents

BAF131610009

Scienceware® biohazard disposal bags

width × H × thickness 610 mm × 30 in. (760 mm) × 0.04 mm (1.5 mil), translucent polypropylene, pack of 100 ea

Synonym(s):

Scienceware® biohazard disposal bags

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.35

material

translucent polypropylene

feature

label (With Biohazard Warning Label and Sterilization Indicator Patch)
Fits waste can 7.3 or 12 gallon

packaging

pack of 100 ea

manufacturer/tradename

Bel-Art F13161-0009

width × H × thickness

610 mm × 30 in. (760 mm) × 0.04 mm (1.5 mil)

Looking for similar products? Visit Product Comparison Guide

General description

Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.

Features and Benefits

  • Transparent polypropylene; 0.03mm (1.5 mil) thick
  • Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
  • High temperature autoclavable at a maximum of 135?C (275?F)
  • Plain, non-printed bags can also be used as conventional trash can liners
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • Bags with biohazard warning label contain standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish

  • Transparent polypropylene; 0.03mm (1.5 mil) thick
  • Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • Plain, non-printed bags can also be used as conventional trash can liners
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minute
  • Bags with biohazard warning label contain standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service