Key Documents
BAF131600009
Scienceware® biohazard disposal bags
width × H × thickness 300 mm (12 in.) × 24 in. (610 mm) × 1.5 mil (0.04 mm), pack of 100 ea, translucent polypropylene
Synonym(s):
Scienceware® biohazard disposal bags
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
translucent polypropylene
feature
label (With Biohazard Warning Label and Sterilization Indicator Patch)
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art F13160-0009
capacity
3.8-11.4 L (1-3 gal)
width × H × thickness
300 mm (12 in.) × 24 in. (610 mm) × 1.5 mil (0.04 mm)
General description
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
Features and Benefits
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
  • Transparent polypropylene; 0.03mm (1.5 mil) thick
  • Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
  • High temperature autoclavable at a maximum of 135?C (275?F)
  • Plain, non-printed bags can also be used as conventional trash can liners
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • Bags with biohazard warning label contain standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
  • Transparent polypropylene; 0.03mm (1.5 mil) thick
  • Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • Plain, non-printed bags can also be used as conventional trash can liners
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • Bags with biohazard warning label contain standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
Legal Information
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)