B8151
Scienceware® biohazard disposal bags
W × H × thickness 8 1/2 in. (22 cm) × 11 in. (28 cm) × 0.00125 in., red, pack of 100
Synonym(s):
autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
Recommended Products
description
Imprinted in black with Biohazard symbol.
packaging
pack of 100
manufacturer/tradename
Bel-Art F13166-0000
W × H × thickness
8 1/2 in. (22 cm) × 11 in. (28 cm) × 0.00125 in.
color
red
Looking for similar products? Visit Product Comparison Guide
Application
Scienceware® biohazarddisposal bags are suitable for disposing of tips and tubes.
Features and Benefits
- ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests .
- For space-saving disposal in fume hoods or on bench tops, use with Poxygrid® Benchtop Biohazard Bag Holder (H13193-1000) and Cover (F13193-0102).
- Autoclave friendly at 121°C (250°F)
- Possesses a long flap for easy opening
- Made of high molecular weight high-density (HMHD) polyethylene
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service