Skip to Content
Merck
All Photos(1)

Key Documents

B66302

Sigma-Aldrich

N-(2-Bromoethyl)phthalimide

95%

Synonym(s):

1-Bromo-2-phthalimidoethane, 2-(2-Bromoethyl)-1H-isoindole-1,3(2H)-dione, 2-(2-Bromoethyl)isoindole-1,3-dione, 2-(2-Bromoethyl)phthalimide, 2-Phthalimidoethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8BrNO2
CAS Number:
Molecular Weight:
254.08
Beilstein:
148736
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

reaction suitability

reagent type: cross-linking reagent

mp

80-83 °C (lit.)

functional group

bromo

SMILES string

BrCCN1C(C2=CC=CC=C2C1=O)=O

InChI

1S/C10H8BrNO2/c11-5-6-12-9(13)7-3-1-2-4-8(7)10(12)14/h1-4H,5-6H2

InChI key

CHZXTOCAICMPQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean Allen et al.
Journal of controlled release : official journal of the Controlled Release Society, 262, 91-103 (2017-07-25)
Flash nanoprecipitation (FNP) has proven to be a powerful tool for the rapid and scalable assembly of solid-core nanoparticles from block copolymers. The process can be performed using a simple confined impingement jets mixer and provides an efficient and reproducible
Brandi D Sanders et al.
Bioorganic & medicinal chemistry, 17(19), 7031-7041 (2009-09-08)
The sirtuin proteins are broadly conserved NAD(+)-dependent deacetylases that are implicated in diverse biological processes including DNA recombination and repair, transcriptional silencing, longevity, apoptosis, axonal protection, insulin signaling, and fat mobilization. Because of these associations, the identification of small molecule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service