Skip to Content
Merck
All Photos(2)

Key Documents

472654

Sigma-Aldrich

N-Boc-2-aminoacetaldehyde

95%

Synonym(s):

tert-Butyl N-(2-oxoethyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOCH2NHCO2C(CH3)3
CAS Number:
Molecular Weight:
159.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.455 (lit.)

functional group

aldehyde
amine

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)NCC=O

InChI

1S/C7H13NO3/c1-7(2,3)11-6(10)8-4-5-9/h5H,4H2,1-3H3,(H,8,10)

InChI key

ACNRTYKOPZDRCO-UHFFFAOYSA-N

Gene Information

human ... CTSK(1513)

General description

N-Boc-2-aminoacetaldehyde is an organic building block. It reacts with Horner-Wadsworth-Emmons (HWE) reagent to afford γ-aminobutyric acid (GABA)-derived α-keto amide/ester units.

Application

α-Methylenation of this amino aldehyde proceeds in a quick and efficient manner using a recently reported protocol involving formaldehyde and catalysis by either pyrrolidine proprionic acid or the dipeptide L-Pro-β-Ala.
Also used in a three-component synthesis of pyrrolidines involving 1,3-dipolar cycloaddition.
N-Boc-2-aminoacetaldehyde may be employed in the following:
  • As a starting reagent in the total synthesis of (+)-negamycin.
  • Synthesis of (E)-ethyl 4-((tert-butoxycarbonyl)amino)but-2-enoate.
  • Synthesis of 2,2′-bipyridine.
A building block in the synthesis of a protected pyrroloproline.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Turetsky et al.
Scientific reports, 4, 4782-4782 (2014-04-25)
A number of Bruton's tyrosine kinase (BTK) inhibitors are currently in development, yet it has been difficult to visualize BTK expression and pharmacological inhibition in vivo in real time. We synthesized a fluorescent, irreversible BTK binder based on the drug
Anniina Erkkilä et al.
The Journal of organic chemistry, 71(6), 2538-2541 (2006-03-11)
A rapid and extremely convenient method for alpha-methylenation of aldehydes with aqueous formaldehyde is described. Two optimal catalytic systems are presented that allow short reaction times and afford the functionalized products in good to excellent yields (up to 99%) and
Diethyl [3-Cyano-2-Oxo-3-(Triphenylphosphoranylidene) propyl] phosphonate: A Useful Horner-Wadsworth-Emmons Reagent for alpha-Keto (Cyanomethylene)-triphenylphosphoranes from Carbonyl Compounds.
Lee K.
Bull. Korean Chem. Soc., 28(10), 1641-1641 (2007)
Yoshio Hayashi et al.
Chemical communications (Cambridge, England), (20)(20), 2379-2381 (2008-05-14)
Herein, we describe an efficient strategy for the total synthesis of (+)-negamycin using commercially available achiral N-Boc-2-aminoacetaldehyde as starting material with 42% overall yield for a limited number of steps.
Claudia Karnthaler-Benbakka et al.
Angewandte Chemie (International ed. in English), 53(47), 12930-12935 (2014-08-01)
The development of receptor tyrosine-kinase inhibitors (TKIs) was a major step forward in cancer treatment. However, the therapy with TKIs is limited by strong side effects and drug resistance. The aim of this study was the design of novel epidermal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service