A81803
4-Amino-4H-1,2,4-triazole
ReagentPlus®, 99%
Synonym(s):
4H-1,2,4-Triazol-4-amine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C2H4N4
CAS Number:
Molecular Weight:
84.08
Beilstein:
107563
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
product line
ReagentPlus®
Assay
99%
form
crystals
mp
84-86 °C (lit.)
SMILES string
Nn1cnnc1
InChI
1S/C2H4N4/c3-6-1-4-5-2-6/h1-2H,3H2
InChI key
FMCUPJKTGNBGEC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jingtang Li et al.
Journal of chromatography. A, 1559, 178-185 (2018-01-07)
Recently,ionic liquids (ILs) functionalized cyclodextrins (CDs) have attracted more and more attention in the fields of enantioseparation. In this study, a novel IL amino triazolium functionalized β-CD derivative, mono-6-deoxy-6-(4-amino-1,2,4-triazolium)-β-cyclodextrin chloride (4-ATMCDCl), was synthesized for the first time and managed to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service