Skip to Content
Merck
All Photos(2)

Key Documents

258938

Sigma-Aldrich

4-Methyl-4H-1,2,4-triazole-3-thiol

97%

Synonym(s):

3-Mercapto-4-methyl-4H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N3S
CAS Number:
Molecular Weight:
115.16
Beilstein:
110580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

165-169 °C (lit.)

solubility

acetone: soluble 2.5%, clear, colorless to faintly yellow

SMILES string

Cn1cnnc1S

InChI

1S/C3H5N3S/c1-6-2-4-5-3(6)7/h2H,1H3,(H,5,7)

InChI key

AGWWTUWTOBEQFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methyl-4H-1,2,4-triazole-3-thiol reacts with ethyl bromoacetate to yield ethyl[(4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]acetate. Electrochemically anticorrosive behavior of self-assembled monolayers of 4-methyl-4H-1,2,4-triazole-3-thiol on the silver electrode has been studied by electrochemical impedance spectroscopy and polarization measurements.

Application

  • Corrosion study of mild steel in aqueous sulfuric acid solution using 4-methyl-4H-1, 2, 4-Triazole-3-Thiol: Examines the use of 4-Methyl-4H-1,2,4-triazole-3-thiol as a corrosion inhibitor for mild steel in sulfuric acid solutions (MEHMETI and BERISHA, 2017).
  • Dimeric and polymeric mercury (II) complexes containing 4-methyl-1, 2, 4-triazole-3-thiol ligand: Investigates the coordination chemistry of mercury with 4-methyl-4H-1,2,4-triazole-3-thiol, providing insights into its potential applications in materials science (TAHERIHA, GHADERMAZI, and AMANI, 2015).
  • Synthesis, characterization and biological studies of 1, 2, 4-triazole derivatives having piperidine moiety: Focuses on the synthesis and biological evaluation of 1,2,4-triazole derivatives, including those modified with a piperidine group and 4-Methyl-4H-1,2,4-triazole-3-thiol (FAROOQ et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclization of 1-{[(4-Methyl-4H-1, 2, 4-triazol-3-yl) sulfanyl] acetyl} thiosemicarbazides to 1, 2, 4-Triazole and 1, 3, 4-Thiadiazole Derivatives and Their Pharmacological Properties.
Maliszewska-Guz A, et al.
Collection of Czechoslovak Chemical Communications, 70(1), 51-62 (2005)
Electrochemical and in situ SERS spectroelectrochemical investigations of 4-methyl-4H-1, 2, 4-triazole-3-thiol monolayers at a silver electrode.
Sun Y, et al.
Journal of Raman Spectroscopy, 40(9), 1306-1311 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service