Skip to Content
Merck
All Photos(4)

Key Documents

94300

Sigma-Aldrich

Urethane

≥99.0% (GC)

Synonym(s):

Carbamic acid ethyl ester, Ethyl carbamate, Ethylurethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COOC2H5
CAS Number:
Molecular Weight:
89.09
Beilstein:
635810
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

3.07 (vs air)

vapor pressure

10 mmHg ( 77.8 °C)

Assay

≥99.0% (GC)

form

crystals

bp

182-184 °C (lit.)

mp

46-49 °C
48-50 °C (lit.)

functional group

amine

SMILES string

CCOC(N)=O

InChI

1S/C3H7NO2/c1-2-6-3(4)5/h2H2,1H3,(H2,4,5)

InChI key

JOYRKODLDBILNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Khoshgoftar et al.
Journal of biomechanics, 48(8), 1427-1435 (2015-03-15)
Since meniscal geometry affects the cartilage contact pressures, it is essential to carefully define the geometry of the synthetic meniscal implant that we developed. Recently, six independent modes of size- and shape-related geometry variation were identified through 3D statistical shape
C A Maggi et al.
Experientia, 42(5), 531-537 (1986-05-15)
The suitability of urethane anesthesia for physiopharmacological experiments in various systems is briefly reviewed. Urethane anesthesia appears to be suitable for various types of studies on respiratory function and on reflex activation of motility of the urinary bladder and some
J Schlatter et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 28(3), 205-211 (1990-03-01)
Ethyl carbamate is found in fermented foods: bread contains 3-15 ng/g, stone-fruit brandies 200-20,000 ng/g, and about one-third of table-wine samples analysed contained more than 10 ng/g. In animals, ethyl carbamate is degraded to CO2, H2O and NH3, with intermediate
C A Maggi et al.
Experientia, 42(2), 109-114 (1986-02-15)
The suitability of urethane anesthesia for physiopharmacological investigations is reviewed. Total dose administered and route of administration are recognized as factors having a great influence on both resting parameters and biological responses to drugs. A peculiar characteristic of urethane is
Elena Amendola et al.
PloS one, 9(5), e91613-e91613 (2014-05-20)
Mutations in cyclin-dependent kinase-like 5 (CDKL5) cause early-onset epileptic encephalopathy, a neurodevelopmental disorder with similarities to Rett Syndrome. Here we describe the physiological, molecular, and behavioral phenotyping of a Cdkl5 conditional knockout mouse model of CDKL5 disorder. Behavioral analysis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service