Skip to Content
Merck
All Photos(1)

Key Documents

755885

Sigma-Aldrich

Urethane acrylate methacrylate resin

Synonym(s):

MAU, U-835

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

Quality Level

refractive index

n20/D 1.487

viscosity

3,000 cP(25 °C)

density

0.995 g/mL at 25 °C

storage temp.

2-8°C

General description

Urethane acrylate methacrylate resin (UDMA) is formed by reacting 2,4,4-trimethylhexamethylene diisocyanate and 2-hydroxyethyl methacrylate (HEMA). It is mainly used as a dental resin with a higher water uptake than that of bisphenol A-glycidyl methacrylate (Bis-GMA).

Application

UDMA may be combined with triethylene glycol dimethacrylate (TEGDMA) as a resin to develop composite blocks, which can be used for dental computer aided design applications. It may also be used to decrease the water sorption and increase the mechanical strength of the dental composites.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>399.2 °F

Flash Point(C)

> 204 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Properties of experimental urethane dimethacrylate-based dental resin composite blocks obtained via thermo-polymerization under high pressure.
Nguyen J, et al.
Dental Materials : Official Publication of the Academy of Dental Materials, 29(5), 535-541 (2013)
Water ageing of urethane dimethacrylate networks.
Pomes B, et al.
Polymer Degradation and Stability, 29(5), 535-541 (2018)
Synthesis and evaluation of modified urethane dimethacrylate resins with reduced water sorption and solubility.
Kerby RE, et al.
Dental Materials : Official Publication of the Academy of Dental Materials, 25(3), 302-313 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service