Skip to Content
Merck
All Photos(1)

Key Documents

P1754

Sigma-Aldrich

TWEEN® 80

viscous liquid

Synonym(s):

TWEEN® 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

biological source

Porcine (tallow)
bovine (tallow)

Quality Level

description

non-ionic

form

viscous liquid

mol wt

micellar avg mol wt 79,000
average mol wt 1310

composition

Oleic acid, ≥58.0% (balance primarily linoleic, palmitic, and stearic acids)

aggregation number

60

CMC

0.012 mM (20-25°C)

viscosity

400-620 mPa.s

transition temp

cloud point 65 °C

solubility

water: soluble, clear to slightly hazy, colorless to yellow

density

1.064 g/cm3

HLB

15

application(s)

cell analysis

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tween® 80 is a non-ionic detergent and surfactant, classified as an ester of sorbitan and oleic acid. It is a versatile compound commonly employed in various biological applications, including cell lysis, nuclei isolation, and cell fractionation. Additionally, Tween® 80 finds use in stabilizing proteins and facilitating protein membrane studies. Its effectiveness stems from its membership in the polysorbate group, characterized by the combination of sorbitol and sorbitane oleic esters with polyethylene glycol chains. These structural features contribute to Tween® 80′s emulsifying capabilities and a broad range of applications.

Application

Non-ionic detergent used for selective protein extraction and isolation of nuclei from mammalian cell lines.
TWEEN® 80 has been used in drug vehicle solution for specific injections in mice and rats. It has been used for the determination of esterase activity in bacterial cells.

Biochem/physiol Actions

TWEEN® 80 is a polysorbate surfactant which helps in stabilizing proteins and in protein membrane studies. It is regularly used for the extraction of membrane proteins and isolation of nuclei from cells.

Features and Benefits

  • Excellent wetting and dispersing agent
  • Highly versatile surfactant for your Cell Biology and Biochemical research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zandra Gerdes et al.
PloS one, 14(2), e0205378-e0205378 (2019-02-20)
The role of microplastic (MP) as a carrier of persistent organic pollutants (POPs) to aquatic organisms has been a topic of debate. However, the reverse POP transport can occur if relative contaminant concentrations are higher in the organism than in
HU-210 shows higher efficacy and potency than morphine after intrathecal administration in the mouse formalin test.
Guhring H et al.
European Journal of Pharmacology, 429, 127-127 (2001)
V Ryu et al.
Neuroscience, 167(4), 1227-1238 (2010-03-04)
Previous reports show that vagal afferent innervation of the stomach eventually regenerates from surviving nodose ganglion (NG) neurons after subdiaphragmatic vagotomy. Systemic capsaicin treatment destroys gastric vagal afferent neurons expressing vanilloid receptor 1 (VR1). However, it is not known whether
Esther Visser et al.
Science advances, 6(19), eaax7060-eaax7060 (2020-06-05)
Alcohol use disorder is characterized by a high risk of relapse during periods of abstinence. Relapse is often triggered by retrieval of persistent alcohol memories upon exposure to alcohol-associated environmental cues, but little is known about the neuronal circuitry that
Yee-Kuen Yam et al.
Frontiers in microbiology, 11, 359-359 (2020-03-21)
Persistence of infection despite extensive chemotherapy with antibiotics displaying low MICs is a hallmark of lung disease caused by Mycobacterium abscessus (Mab). Thus, the classical MIC assay is a poor predictor of clinical outcome. Discovery of more efficacious antibiotics requires

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service