Skip to Content
Merck
All Photos(2)

Key Documents

934690

Sigma-Aldrich

Graphite

greener alternative

electrode sheet, copper substrate, size 5 in. × 10 in.

Synonym(s):

Carbon anode, Graphite anode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.21
Pricing and availability is not currently available.

material

copper substrate (current collector)

Quality Level

grade

battery grade

description

Description/Nominal Voltage: 0.1 V vs Li/Li+

Assay

≥98% (active material characteristic)

composition

Active material loading 6.5 mg/cm2 ± 5%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

size

10 μm , copper current collector
5 in. × 10 in.
60 μm , excluding current collector

particle size

15-20 μm (active material characteristic)

capacity

≥370 mAh/g (Nominal Discharge)

application(s)

battery manufacturing

greener alternative category

General description

Graphite electrode sheet is a ready-to-use graphite anode film, cast single-sided on 10 µm thick copper foil current collectors. The sheets are 5 inches x 10 inches (127 mm x 254 mm). The composition is 90% graphite, 5% Poly(vinylidene fluoride) [PVDF] and 5% Carbon Black. PVDF acts as binder due to its thermal and electrochemical stability.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Our graphite electrode sheet is ready-to-use as an anode in rechargeable batteries, especially lithium-ion batteries and potassium-ion batteries. The primary advantages of graphite electrodes are a low insertion/deinsertion potential (0.1V vs. Li/Li+) and a high capacity, exceeding 340 mAh/g, coupled with stable cycling performance from the lack of dendrite formation. Our graphite electrodes have a high active material loading (90%) and achieve high areal capacity (2.4 mAh/cm2), allowing them to operate even at high charge currents like 5C. These electrodes offer excellent, consistent performance making them useful for research as a benchmark and counter electrode for experimental studies of cathode and electrolyte optimization as well as mechanistic in operando studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graphite as anode materials: Fundamental mechanism, recent progress and advances
Zhang H, et al.
Energy Storage Materials, 36, 147-170 (2021)
Graphite as anode materials: Fundamental mechanism, recent progress and advances
Zhang Hao, et al
Energy Storage Materials, 36, 147-170 (2021)
Recent progress of electrode materials cooperated with potassium bis(fluorosulfonyl)imide?containing electrolyte for K-ion batteries
Xie, Junpeng, et al.
Materials Today, 6, 100035-100035 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service