Skip to Content
Merck
All Photos(2)

Key Documents

934054

Sigma-Aldrich

Aluminum oxide

greener alternative

gamma phase, nanopowder, 5 nm avg. part. size, 99.95% trace metals basis

Synonym(s):

γ-Al2O3, Cubic aluminium oxide, Gamma-alumina, Gamma-aluminum(III) oxide, Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
MDL number:
UNSPSC Code:
12352311
NACRES:
NA.21

Quality Level

Assay

99.95% trace metals basis

form

nanopowder

composition

Al2O3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

avg. part. size

5 nm

particle size

5 nm

mp

2040 °C (lit.)

bulk density

0.18 g/cm3

application(s)

battery manufacturing

greener alternative category

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gamma-phase Al2O3 is one of the several crystalline phases of aluminum oxide, also known as alumina. It has a cubic crystal structure with a high degree of symmetry, characterized by a closely packed arrangement of oxygen and aluminum ions, and tends to be highly porous. Our aluminum oxide nanopowder in the gamma phase is a high-purity product with 99.95% purity and an average particle size of 5 nm. This nanopowder is characterized by its high surface area, excellent dielectric properties, and resistance to corrosion and wear. Our aluminum oxide nanopowder is produced using advanced manufacturing techniques, ensuring uniform particle size and high purity.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product has been enhanced for energy efficency. Click here for more information.

Application

Our gamma phase alumina nanopowder is useful in a variety of applications. Its high purity and small particle size make it an excellent catalyst and catalyst-support for chemical reactions, including dehydration, isomerization, hydrogenation, oxidation, reforming, and Fischer-Tropsch synthesis. Because of its porosity, it has many uses in pharmaceutical and industrial manufacturing processes as an adsorbent and desiccating agent. In addition, gamma-phase Al2O3 is a widely used material in the field of electronics due to its high dielectric constant, low dielectric loss, and excellent electrical insulation properties. Its ability to store electrical charge, retain its properties over a wide range of frequencies, and withstand high voltages make it an ideal choice for use in a wide range of electronic components, including capacitors, insulators, transformers, and high-frequency and high-voltage applications. The nanopowder′s hardness and stability make it suitable for use in wear-resistant coatings, and its resistance to corrosion and oxidation makes it a reliable material for high-temperature environments.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomedical applications of aluminium oxide nanoparticles
Hassanpour P, et al.
Micro and Nano Systems Letters, 13, 1227 ? 1231-1227 ? 1231 (2018)
Chitosan/Gamma-Alumina/Fe3O4@5-FU Nanostructures as Promising Nanocarriers: Physiochemical Characterization and Toxicity Activity
Ajalli N, et al.
Molecules (Basel), 27, 5369-5369 (2022)
Khadije Fasihi et al.
Tissue & cell, 76, 101755-101755 (2022-02-28)
5-Fluorouracil (5-FU) is being used in the treatment of several malignancies, but side effects are often reported and include: diarrhea, vomiting, nausea, poor appetite, watery eyes, and photophobia. We have developed and tested the cytotoxic activity of nanocrystalline powder of
New sol-gel derived aluminum oxide-ibuprofen nanocomposite as a controlled releasing medication
Tarlani A, et al.
Nanomedicine research journal, 2, 28-35 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service