931837
(4R,4′R,5S,5′S)-2,2′-Cyclopropylidenebis[4,5-dihydro-4,5-diphenyloxazole]
≥95%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C33H28N2O2
CAS Number:
Molecular Weight:
484.59
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
form
powder
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
SMILES string
N1=C(O[C@H]([C@H]1c7ccccc7)c6ccccc6)C5(CC5)C2=N[C@@H]([C@@H](O2)c4ccccc4)c3ccccc3
InChI key
UAXGHJCFHMQPBJ-XAZDILKDSA-N
Related Categories
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.
Application
(4R,4′R,5S,5′S)-2,2′-Cyclopropylidenebis[4,5-dihydro-4,5-diphenyloxazole] is a bisoxazoline (BOX) ligand for asymmetric catalysis. The copper-ligand complex has been shown to catalyze reactions of symmetric diaziridines with enol diazo compounds.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Catalytic Desymmetric Cycloaddition of Diaziridines with Metalloenolcarbenes: The Role of Donor-Acceptor Cyclopropenes
Zheng H, et al.
Angewandte Chemie (International Edition in English), 58, 12502-12506 (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service