464155
[3aR-[2(3′aR*,8′aS*),3′aβ,8′aβ]]-(+)-2,2′-Methylenebis[3a,8a-dihydro-8H-indeno[1,2-d]oxazole]
98%
About This Item
Recommended Products
Assay
98%
optical activity
[α]22/D +353°, c = 3.7 in chloroform
mp
204-206 °C (lit.)
functional group
ether
SMILES string
C1[C@@H]2OC(CC3=N[C@H]4[C@@H](Cc5ccccc45)O3)=N[C@H]2c6ccccc16
InChI
1S/C21H18N2O2/c1-3-7-14-12(5-1)9-16-20(14)22-18(24-16)11-19-23-21-15-8-4-2-6-13(15)10-17(21)25-19/h1-8,16-17,20-21H,9-11H2/t16-,17-,20+,21+/m0/s1
InChI key
BDHSVQLSNIGJNC-ZCLUNYJNSA-N
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.
BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.
BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.
BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service