900966
PFN-DOF
Mw >10,000
Synonym(s):
Poly[(9,9-bis(3′-(N,N-dimethylamino)propyl)-2,7-fluorene)-alt-2,7-(9,9-dioctylfluorene)]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
Quality Level
mol wt
Mw >10,000
solubility
organic solvents: soluble (CH2Cl2, CHCl3, Toluene)
λmax
378 nm in dichloromethane
fluorescence
λem 414 nm in dichloromethane (PL)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Phosphorescent host material. Polymer for use in electron transport layer in OPV and OLED applications.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Novel Electroluminescent Conjugated Polyelectrolytes Based on Polyfluorene.
Huang F, et al.
Chemistry of Materials, 16(4), 708-716 (2004)
Changfeng Han et al.
ACS applied materials & interfaces, 8(5), 3301-3307 (2016-01-13)
A highly efficient inverted polymer solar cell (PSC) has been successfully demonstrated by using a ZnO nanoparticle (NP) and poly[(9,9-bis(3'-(N,N-dimethylamino)propyl)-2,7-fluorene)-alt-2,7-(9,9-dioctyfluorene)] (PFN) bilayer structure as an effective electron collecting layer. This ZnO/PFN bilayer structure is designed to combine the advantages of
Fei Huang et al.
Journal of the American Chemical Society, 126(31), 9845-9853 (2004-08-05)
A series of aminoalkyl-substituted polyfluorene copolymers with benzothiadiazole (BTDZ) of different content were synthesized by Suzuki coupling reaction, and their quaternized ammonium polyelectrolyte derivatives were obtained through a postpolymerization treatment on the terminal amino groups. Copolymers are soluble in environmentally
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service