Skip to Content
Merck
All Photos(2)

Key Documents

760986

Sigma-Aldrich

Lithium nickel cobalt oxide

greener alternative

powder, <0.5 μm particle size, >98%

Synonym(s):

LNCO, Lithium cobalt nickel oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNi0.8Co0.2O2
CAS Number:
UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

Quality Level

Assay

>98%

form

powder

mol wt

Mw 97.68 g/mol

composition

LiNi0.8Co0.2O2

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

<0.5 μm

mp

>1000 °C (lit.)

application(s)

battery manufacturing

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

LNCO is a lamellar oxide used as a positive electrode material in Li batteries exhibiting good electrochemical charge-discharge behavior. The high reversibility of the lithium intercalation process, due to the strictly 2D character of the LNCO material, the high cell voltage (but not too high due to the position of the Ni4+/Ni3+ couple vs the Co4+/Co3+ couple), and the presence of a relatively small cobalt content, makes this material a very promising one for lithium battery applications.

Legal Information

Product of Engi-Mat Co.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saadoune, I.; et al.
Journal of Solid State Chemistry, 136, 8-8 (1998)
Andersson, A. M.; et al.
Journal of the Electrochemical Society, 149, A1358-A1358 (2002)
Ganter, M. J.; et al.
Electrochimica Acta, 56, 7272-7272 (2011)

Articles

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service