Skip to Content
Merck
All Photos(1)

Key Documents

806781

Sigma-Aldrich

TPBi

Synonym(s):

2,2′,2"-(1,3,5-Benzinetriyl)-tris(1-phenyl-1-H-benzimidazole)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H30N6
CAS Number:
Molecular Weight:
654.76
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.5% (HPLC)

Quality Level

form

powder

mp

272-277 °C

Orbital energy

HOMO 6.2 eV 
LUMO 2.7 eV 

OLED Device Performance

ITO/MoO3/NPB/Cz-BTPE/TPBi/LiF/Al (ref 2)

  • Color: blue
  • Max. Luminance: 9911 Cd/m2
  • Max. EQE: 1.9 %
  • Turn-On Voltage: 4.9 V

ITO/MoO3/NPB/Ph-BTPE/TPBi/LiF/Al
  • Color: blue
  • Max. Luminance: 6497 Cd/m2
  • Max. EQE: 1.9 %
  • Turn-On Voltage: 5.3 V

ITO/MoO3/NPB/isopro-BTPE/TPBi/LiF/Al(ref 2)
  • Color: blue
  • Max. Luminance: 1127 Cd/m2
  • Max. EQE: 1.7 %
  • Turn-On Voltage: 5.3 V

ITO/MoO3/NPB/methyl-BTPE/TPBi/LiF/Al (ref 2)
  • Color: blue
  • Max. Luminance: 1976 Cd/m2
  • Max. EQE: 1.3 %
  • Turn-On Voltage: 5.3 V

SMILES string

C1(C2=NC(C=CC=C3)=C3N2C4=CC=CC=C4)=CC(C5=NC(C=CC=C6)=C6N5C7=CC=CC=C7)=CC(C8=NC(C=CC=C9)=C9N8C%10=CC=CC=C%10)=C1

InChI

1S/C45H30N6/c1-4-16-34(17-5-1)49-40-25-13-10-22-37(40)46-43(49)31-28-32(44-47-38-23-11-14-26-41(38)50(44)35-18-6-2-7-19-35)30-33(29-31)45-48-39-24-12-15-27-42(39)51(45)36-20-8-3-9-21-36/h1-30H

InChI key

GEQBRULPNIVQPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TPBi is an organic semiconductor that displays high electron mobility (3.3×10−5 cm2 V−1 s−1) and high glass transition temperature. It is widely used as an electron transport layer(ETL) in QLEDs and OLEDs, which is a cathode buffer layer in organic solar cells.

The increase in power conversion efficiency of the device on the addition of TBPi is attributed to the following reasons:
  • Preventing exciton from quenching at the acceptor/cathode interface.
  • Acting as an optical spacer.
  • Forming a passivating layer.

Application

TPBi can be used to fabricate an electron transport layer for organic-inorganic hybrid perovskite LEDs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Study on Electron Transport Characterization in TPBi Thin Films and OLED Application
Yanping Wang, et al.
The Journal of Physical Chemistry C, 125, 16753-16758 (2021)
Organic photovoltaic cells based on TPBi as a cathode buffer layer
Junsheng Yu, et al.
Solar Energy Materials and Solar Cells, 95, 664-668 (2011)
Employing ~100% Excitons in OLEDs by Utilizing a Fluorescent Molecule with Hybridized Local and Charge-Transfer Excited State
Weijun L, et al.
Advances in Functional Materials, 24, 1609-1614 (2014)
High-efficiency deep-blue organic light-emitting diodes based on a thermally activated delayed fluorescence emitter
Shuanghong W, et al.
Journal of Material Chemistry C, 2, 421-424 (2014)
Largely blue-shifted emission through minor structural modifications: molecular design, synthesis, aggregation-induced emission and deep-blue OLED application
Jing H, et al.
Chemical Communications (Cambridge, England), 50, 2136-2138 (2014)

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service