Skip to Content
Merck
All Photos(2)

Key Documents

900928

Sigma-Aldrich

Liq

greener alternative

sublimed

Synonym(s):

(8-Hydroxyquinolinato)lithium, (8-Quinolinolato)lithium, 8-Hydroxyquinoline lithium, 8-Quinolinol lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6NOLi
CAS Number:
Molecular Weight:
151.09
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

Absorption (UV): 261 nm (in THF)
Emission (PL): 331 nm (in THF)

Quality Level

Assay

>99.5% (sublimed, HPLC)

form

powder
sublimed

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

363-368 °C

greener alternative category

InChI

1S/C9H7NO.Li/c11-8-5-1-3-7-4-2-6-10-9(7)8;/h1-6,11H;/q;+1/p-1

InChI key

FQHFBFXXYOQXMN-UHFFFAOYSA-M

General description

Electron transport and hole blocking material in organic light emitting diodes (OLED).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Electron transport organic materials have the chemical functionality to transport electrons and are used as electron transport layer in OLED devices. Click here for more information.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of 2,3,6,7-tetraarylbenzo[1,2-b:4,5-b']difurans as hole-transporting material.
Hayato Tsuji et al.
Journal of the American Chemical Society, 129(39), 11902-11903 (2007-09-14)
Lithium-Quinolate Complexes as Emitter and Interface Materials in Organic Light-Emitting Diodes.
Schmitz C, et al.
Chemistry of Materials, 12(10), 3012-3019 (2000)
Extremely efficient flexible organic light-emitting diodes with modified graphene anode.
Han TH, et al.
Nature Photonics, 6(2), 105-110 (2012)

Articles

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service