Skip to Content
Merck
All Photos(1)

Key Documents

338001

Sigma-Aldrich

Zirconium(IV) acetylacetonate

97%

Synonym(s):

2,4-Pentanedione zirconium(IV) derivative, Tetrakis(2,4-pentanedionato)zirconium(IV), Zirconium(IV) 2,4-pentanedionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(C5H7O2)4
CAS Number:
Molecular Weight:
487.66
Beilstein:
4160373
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

reaction suitability

core: zirconium
reagent type: catalyst

SMILES string

CC(=O)\C=C(\C)O[Zr](O\C(C)=C/C(C)=O)(O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/4C5H8O2.Zr/c4*1-4(6)3-5(2)7;/h4*3,6H,1-2H3;/q;;;;+4/p-4/b4*4-3-;

InChI key

FPFOSIXCIBGKOH-MTOQALJVSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Zirconium(IV) acetylacetonate is a white crystalline organometallic solid widely used as a sol-gel precursor to synthesize Zirconium-based nanomaterials and thin films. It also finds application in the field of catalysis and polymer synthesis.

Application

Zirconium(IV) acetylacetonate can be used:
  • As a sol-gel precursor to synthesize zirconia nanoparticles with photocatalytic activity.
  • As an additive to improve the performance of the electron transport layer in perovskite solar cells.
  • To synthesize as-prepared zirconium acetylacetonate (a-ZrAcac) film as a cathode buffer layer in polymer solar cells. It exhibited an average power conversion efficiency (PCE) of 8.75% which is significantly improved than that of the devices with traditional Ca/Al cathode.
  • To synthesize high-κ zirconium oxide gate dielectric for Organic Field-Effect Transistors at low voltage operation.
  • To fabricate the conductive interlayers for quantum dot light-emitting devices(QLEDs), which enhances quantum efficiency, power efficiency, and longer operational lifetimes.
  • As a ring-opening initiator to synthesize biodegradable polyacids.
  • As a catalyst to synthesize polylactide terpolymers.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of polylactide terpolymers P(LLA-TMC-GA) catalyzed by zirconium(IV)acetylacetonate
Shaohua Wen, et al
Applied Organometallic Chemistry, 32, e4177-e4177 (2018)
Improving the TiO2 electron transport layer in perovskite solar cells using acetylacetonate-based additives
Hsin-Hua Wang, et al.
Journal of Materials Chemistry, 3, 9108-9115 (2015)
High performance polymer solar cells with as-prepared zirconium acetylacetonate film as cathode buffer layer
Zhan'ao Tan, et al.
Scientific Reports, 4, 4691-4691 (2014)
Zirconium (IV) acetylacetonate: Ring-opening initiator mediating one-step synthesis of biodegradable polyacids
Piotr Dobrzy'nski, et al.
Advances in Polymer Technology, 2019-2019 (2019)
Room-temperature fabrication of ultrathin oxide gate dielectrics for low-voltage operation of organic field-effect transistors
Young Min Park, et al.
Advanced Materials, 23, 971-974 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service