Skip to Content
Merck
All Photos(1)

Key Documents

900540

Sigma-Aldrich

Ir(dFppy)3

Synonym(s):

tris[3,5-difluoro-2-(2-pyridinyl)phenyl]-Iridium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H18F6IrN3
CAS Number:
Molecular Weight:
762.72
UNSPSC Code:
12161600
NACRES:
NA.22
Pricing and availability is not currently available.

form

powder or crystals

Quality Level

reaction suitability

core: iridium
reaction type: Photocatalysis
reagent type: catalyst

mp

401 °C

photocatalyst activation

450 nm

SMILES string

FC1=CC([Ir]C2=C(C3=NC=CC=C3)C(F)=CC(F)=C2)=C(C4=NC=CC=C4)C(F)=C1.FC5=CC(F)=CC=C5C6=NC=CC=C6

InChI

1S/3C11H6F2N.Ir/c3*12-8-4-5-9(10(13)7-8)11-3-1-2-6-14-11;/h3*1-4,6-7H;

InChI key

GFSFWLGQTIKUOK-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
900539900570901409
form

powder or crystals

form

powder or crystals

form

powder or crystals

form

solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

mp

401 °C

mp

372.5 °C

mp

370.93 °C

mp

-

photocatalyst activation

450 nm

photocatalyst activation

430 nm

photocatalyst activation

465 nm

photocatalyst activation

450 nm

Application

Ir(dFppy)3 is a photocatalyst that readily facilitates the decarboxylative arylation of α amino acids using visible light.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service