Skip to Content
Merck
All Photos(3)

Key Documents

904767

Sigma-Aldrich

[Ru(phen)3]Cl2

≥95%

Synonym(s):

Dichlorotris(1,10-phenanthroline)ruthenium(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H24Cl2N6Ru
CAS Number:
Molecular Weight:
712.59
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Photocatalysis

mp

>300 °C

InChI

1S/3C12H8N2.2ClH.Ru/c3*1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;;/h3*1-8H;2*1H;/q;;;;;+2/p-2

InChI key

UWXWBVKIJZGXQL-UHFFFAOYSA-L

Application

[Ru(phen)3]Cl2 is a cyclometalated ruthenium complex that can be used in visible light mediated photocatalytic organic transformations.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Chen et al.
Chemical communications (Cambridge, England), 54(50), 6780-6783 (2018-02-06)
A visible light-driven photocatalytic generation of sulfonamidyl radicals, and application to intramolecular alkene hydroamination, has been accomplished, providing a mild and efficient approach to various functionalized isoxazolidines. The success of this protocol is based on the strategy of oxidative deprotonation
Tanmay Chatterjee et al.
The Journal of organic chemistry, 81(16), 6995-7000 (2016-06-22)
A simple and practical method for the synthesis of substituted oxazoles has been developed using readily available α-bromoketones and benzylamines by visible-light photocatalysis at room temperature. The process, which requires 1 mol % of [Ru(bpy)3]Cl2 photocatalyst with K3PO4 and CCl3Br
Adam Noble et al.
Angewandte Chemie (International ed. in English), 57(8), 2155-2159 (2018-01-10)
The synthesis of alkyl boronic esters by direct decarboxylative radical addition of carboxylic acids to vinyl boronic esters is described. The reaction proceeds under mild photoredox catalysis and involves an unprecedented single-electron reduction of an α-boryl radical intermediate to the
Chelsea A Huff et al.
The Journal of organic chemistry, 81(16), 6980-6987 (2016-06-18)
We report the development of a method for room-temperature C-H hydroxymethylation of heteroarenes. A key enabling advance in this work was achieved by implementing visible light photoredox catalysis that proved to be applicable to many classes of heteroarenes and tolerant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service