Skip to Content
Merck
All Photos(2)

Key Documents

901368

Sigma-Aldrich

[Ir(dFppy)2(dtbbpy)]PF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H36F10IrN4P
CAS Number:
Molecular Weight:
985.92
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.23

form

powder or crystals

Quality Level

reaction suitability

core: iridium
reagent type: catalyst
reaction type: Photocatalysis

mp

>300 °C

photocatalyst activation

465 nm

SMILES string

[Ir+3]5([n+]6c(cc(cc6)C(C)(C)C)c7[n+]5ccc(c7)C(C)(C)C)(c3c(c(cc(c3)F)F)c4ncccc4)c1c(c(cc(c1)F)F)c2ncccc2.F[P-](F)(F)(F)(F)F

InChI

1S/C18H24N2.2C11H6F2N.F6P.Ir/c1-17(2,3)13-7-9-19-15(11-13)16-12-14(8-10-20-16)18(4,5)6;2*12-8-4-5-9(10(13)7-8)11-3-1-2-6-14-11;1-7(2,3,4,5)6;/h7-12H,1-6H3;2*1-4,6-7H;;/q;;;-1;+5

InChI key

QHIBKCJQTBUWRW-UHFFFAOYSA-N

Application

This photocatalyst readily facilitates alkynylation of carboxylic acids as well as the decarboxylative cross-coupling of oxalates via metallaphotoredox catalysis.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaheng Zhang et al.
Journal of the American Chemical Society (2016-10-11)
Alkyl oxalates, prepared from their corresponding alcohols, are engaged for the first time as carbon radical fragments in metallaphotoredox catalysis. In this report, we demonstrate that alcohols, native organic functional groups, can be readily activated with simple oxalyl chloride to
Timothy M Monos et al.
The Journal of organic chemistry, 81(16), 6988-6994 (2016-06-16)
We report a rapid, one-pot, operationally simple, and scalable preparation of valuable cationic heteroleptic iridium(III) polypyridyl photosensitizers. This method takes advantage of two consecutive microwave irradiation steps in the same reactor vial, avoiding the need for additional reaction purifications. A
Franck Le Vaillant et al.
Angewandte Chemie (International ed. in English), 54(38), 11200-11204 (2015-07-28)
Alkynes are used as building blocks in synthetic and medicinal chemistry, chemical biology, and materials science. Therefore, efficient methods for their synthesis are the subject of intensive research. Herein, we report the direct synthesis of alkynes from readily available carboxylic
Strongly Blue Luminescent Cationic Iridium(III) Complexes with an Electron-Rich Ancillary Ligand: Evaluation of Their Optoelectronic and Electrochemiluminescence Properties.
Ladouceur S,et al.
European Journal of Inorganic Chemistry, 2013, 5329-5343 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service