Skip to Content
Merck
All Photos(1)

Key Documents

87993

Sigma-Aldrich

Tetraoctylammonium hydroxide solution

~20% in methanol (T)

Synonym(s):

Tetra-N-octyl ammonium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7]4N(OH)
CAS Number:
Molecular Weight:
483.90
Beilstein:
7323584
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

~20% in methanol (T)

anion traces

bromide (Br-): ≤500 mg/kg

functional group

amine

storage temp.

2-8°C

SMILES string

[OH-].CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC

InChI

1S/C32H68N.H2O/c1-5-9-13-17-21-25-29-33(30-26-22-18-14-10-6-2,31-27-23-19-15-11-7-3)32-28-24-20-16-12-8-4;/h5-32H2,1-4H3;1H2/q+1;/p-1

InChI key

DCFYRBLFVWYBIJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tetraoctylammonium hydroxide solution can be used as:
  • A lipophilic base in the study of its effect on optical pCO2 sensors.
  • A lipophilic organic base in the preparation of nanobeads, which are used as sensors for the detection of acidic and basic gases.
  • A starting material for the preparation of ionic liquid named tetraoctylammonium 2-methyl-1-naphthoate, which is used in the extraction of 2,3-butanediol from H2O.

Other Notes

Very lipophilic phase-transfer agent; employed in a carbon dioxide optical sensor

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Precipitation as a simple and versatile method for preparation of optical nanochemosensors
Borisov SM, et al.
Talanta, 79(5), 1322-1330 (2009)
The influence of the lipophilic base in solid state optical pCO2 sensors
Schroder CR and Klimant I
Sensors and Actuators B, Chemical, 107(2), 572-579 (2005)
A. Mills et al.
Analytical Chemistry, 64, 1383-1383 (1992)
(Liquid+ liquid) equilibrium data for the separation of 2, 3-butanediol from aqueous streams using tetraoctyl ammonium 2-methyl-1-naphthoate
Garcia-Chavez LY, et al.
The Journal of Chemical Thermodynamics, 55, 85-91 (2012)
J P Mendes et al.
Sensors (Basel, Switzerland), 19(24) (2019-12-19)
A sensing configuration for the real-time monitoring, detection, and quantification of dissolved carbon dioxide (dCO2) was developed for aquaculture and other applications in freshwater and saline water. A chemical sensing membrane, based on a colorimetric indicator, is combined with multimode

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service