857319
Chloramine-T hydrate
95%
Synonym(s):
N-Chloro-p-toluenesulfonamide sodium salt hydrate
About This Item
Recommended Products
Quality Level
Assay
95%
form
solid
reaction suitability
reagent type: oxidant
mp
167-170 °C (dec.) (lit.)
SMILES string
O.Cc1ccc(cc1)S(=O)(=O)N([Na])Cl
InChI
1S/C7H7ClNO2S.Na.H2O/c1-6-2-4-7(5-3-6)12(10,11)9-8;;/h2-5H,1H3;;1H2/q-1;+1;
InChI key
BXSDMMPOQRECKB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Preparation of factor Xa inhibitors as novel anticoagulants
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service