Skip to Content
Merck
All Photos(3)

Key Documents

806056

Sigma-Aldrich

Guanidinium iodide

greener alternative

≥99%

Synonym(s):

Aminoformamidine hydriode, Diaminomethaniminium iodide, Greatcell Solar®, Guanidine hydriodide, Guanidine monohydroiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH6IN3
CAS Number:
Molecular Weight:
186.98
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

Quality Level

Assay

≥99%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

194-199 °C

greener alternative category

SMILES string

[nH2+]c([nH])[nH].[I-]

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Guanidinium iodide (GI) belongs to the class of guanidinium salts that crystallize in polar symmetry. It can be used as a passivating material for enhancing the grain boundaries and improving the open-circuit voltage. It can further be used in the fabrication of polymeric solar cells (PSCs).
The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of formamidinium and guanidinium lead tri-iodide powders as precursors for solar cells
Dimesso L, et al.
Materials Science and Engineering, B, 204, 27-33 (2016)
Perovskite solar cells yielding reproducible photovoltage of 1.20 V
Alharbi EA, et al.
Research (Washington, D.C.), 2019, 8474698-8474698 (2019)
Origin of spontaneous polarization and reconstructive phase transition in guanidinium iodide
Szafranski M and Jarek M
CrystEngComm, 15(23), 4617-4623 (2013)
Nam Joong Jeon et al.
Nature, 517(7535), 476-480 (2015-01-07)
Of the many materials and methodologies aimed at producing low-cost, efficient photovoltaic cells, inorganic-organic lead halide perovskite materials appear particularly promising for next-generation solar devices owing to their high power conversion efficiency. The highest efficiencies reported for perovskite solar cells
Zhi-Kuang Tan et al.
Nature nanotechnology, 9(9), 687-692 (2014-08-05)
Solid-state light-emitting devices based on direct-bandgap semiconductors have, over the past two decades, been utilized as energy-efficient sources of lighting. However, fabrication of these devices typically relies on expensive high-temperature and high-vacuum processes, rendering them uneconomical for use in large-area

Articles

A brief tutorial on alternative energy materials for advanced batteries and fuel cells, as well as high-purity inorganics, conducting polymers, and electrolytes.

A brief tutorial on alternative energy materials for advanced batteries and fuel cells, as well as high-purity inorganics, conducting polymers, and electrolytes.

A brief tutorial on alternative energy materials for advanced batteries and fuel cells, as well as high-purity inorganics, conducting polymers, and electrolytes.

A brief tutorial on alternative energy materials for advanced batteries and fuel cells, as well as high-purity inorganics, conducting polymers, and electrolytes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service