Skip to Content
Merck
All Photos(2)

Key Documents

793493

Sigma-Aldrich

Methylammonium iodide

greener alternative

98%

Synonym(s):

Methanamine hydriodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH2 • HI
CAS Number:
Molecular Weight:
158.97
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

270-280 °C

greener alternative category

SMILES string

CN.I

InChI

1S/CH5N.HI/c1-2;/h2H2,1H3;1H

InChI key

LLWRXQXPJMPHLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methanamine hydriodide is an important precursor for the preparation of perovskite photoactive layers for solar energy conversion.
Methylammonium iodide (MAI) is extensively used as a precursor material for the fabrication of perovskite solar cells. These solar cells offer high conversion efficiencies.
Methylammonium iodide can be used as a precursor in combination with lead iodide to change the morphology of the resulting perovskite materials. Perovskite materials can further be utilized in the fabrication of alternative energy devices such as light emitting diodes (LEDs), and perovskite solar cells (PSCs).

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient planar heterojunction mixed-halide perovskite solar cells deposited via spray-deposition.
Barrows AT, et al.
Energy & Environmental Science, 7(9), 2944-2950 (2014)
Benjamin T Diroll
The journal of physical chemistry letters, 10(18), 5623-5628 (2019-09-11)
Intraband relaxation in polycrystalline films of hybrid perovskites methylammonium lead tribromide and methylammonium lead triiodide are studied by transient absorption spectroscopy from 80 K to >350 K. This temperature range spans the transitions of these materials from the high-temperature cubic
Parameters influencing the deposition of methylammonium lead halide iodide in hole conductor free perovskite-based solar cells.
Cohen Bat-El, et al.
APL Materials, 2(8), 081502-081502 (2014)
Crystallization of a perovskite film for higher performance solar cells by controlling water concentration in methyl ammonium iodide precursor solution
Adhikari N, et al.
Nanoscale, 8(5), 2693-2703 (2016)
Siva Uthayaraj et al.
Materials (Basel, Switzerland), 12(13) (2019-06-30)
This study focuses on employing cuprous iodide (CuI) as a hole-transporting material (HTM) in fabricating highly efficient perovskite solar cells (PSCs). The PSCs were made in air with either CuI or 2,2',7,7'-Tetrakis[N,N-di(4-methoxyphenyl)amino]-9,9'-spirobifluorene (spiro-OMeTAD) as HTMs. A simple and novel pressing

Articles

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

A brief tutorial on alternative energy materials for advanced batteries and fuel cells, as well as high-purity inorganics, conducting polymers, and electrolytes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service