Skip to Content
Merck
All Photos(2)

Key Documents

481688

Sigma-Aldrich

Poly(dimethylsiloxane), bis(3-aminopropyl) terminated

average Mn ~2,500

Synonym(s):

PDMS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)3Si(CH3)2O[Si(CH3)2O]nSi(CH3)2(CH2)3NH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

specific activity

0.6-0.8 meq/g (Amine number)

Quality Level

mol wt

average Mn ~2,500

refractive index

n20/D 1.410

viscosity

50 cSt(lit.)

bp

>200 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)=O.C[Si](C)(O)CCCN

Looking for similar products? Visit Product Comparison Guide

General description

Tandem mass spectrometry (MS/MS) reveals that bis(3-aminopropyl)-PDMS mainly dissociates via intramolecular nucleophilic substitutions at the end groups.

Application

bis(3-aminopropyl) terminated poly(dimethylsiloxane) may be used in the synthesis of dipicolylamidopropylpoly(dimethylsiloxane) (DPPDMS).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal decomposition behavior of cobalt (II)picolyl-terminated poly(dimethylsiloxane)
Kurokowa A, et al.
Journal of Thermal Analysis and Calorimetry, 97, 595-599 null
Thierry Fouquet et al.
Journal of the American Society for Mass Spectrometry, 22(4), 649-658 (2011-04-08)
Ammonium adducts of trimethylsilyl-terminated poly(dimethylsiloxane) (CH(3)-PDMS) produced by electrospray ionization were submitted to collision induced dissociation and revealed a particular MS/MS behavior: the same three main product ions at m/z 221, 295, and 369 were always generated in very similar

Articles

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

See All

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service