794384
Cholesteryl-TEG azide
Synonym(s):
3-(13-azido-5,8,11-trioxa-2-azatridecanoate) cholesterol
About This Item
Recommended Products
form
solid
Quality Level
storage temp.
−20°C
SMILES string
CC(C)CCC[C@@H](C)[C@@]1([H])CC[C@@]2([H])[C@]3([H])CC=C4C[C@@H](OC(NCCOCCOCCOCCN=[N+]=[N-])=O)CC[C@]4(C)[C@@]3([H])CC[C@@]21C
InChI
1S/C36H62N4O5/c1-26(2)7-6-8-27(3)31-11-12-32-30-10-9-28-25-29(13-15-35(28,4)33(30)14-16-36(31,32)5)45-34(41)38-17-19-42-21-23-44-24-22-43-20-18-39-40-37/h9,26-27,29-33H,6-8,10-25H2,1-5H3,(H,38,41)/t27-,29+,30+,31-,32+,33+,35+,36-/m1/s1
InChI key
POWZASPNLQFLJO-MKQVXYPISA-N
Related Categories
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service