Skip to Content
Merck
All Photos(1)

Key Documents

762024

Sigma-Aldrich

Azide-PEG3-biotin conjugate

Synonym(s):

Polyethylene glycol, N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]hexahydro-2-oxo-(3aS,4S,6aR)- 1H-thieno[3,4-d ]imidazole-4-pentanamide, Biotin-PEG3-azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32N6O5S
CAS Number:
Molecular Weight:
444.55
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

SMILES string

O=C1N[C@](CS[C@H]2CCCCC(NCCOCCOCCOCCN=[N+]=[N-])=O)([H])[C@]2([H])N1

InChI

1S/C18H32N6O5S/c19-24-21-6-8-28-10-12-29-11-9-27-7-5-20-16(25)4-2-1-3-15-17-14(13-30-15)22-18(26)23-17/h14-15,17H,1-13H2,(H,20,25)(H2,22,23,26)/t14-,15-,17-/m1/s1

InChI key

ZWFOOMQCYIGZBE-BFYDXBDKSA-N

Application

Azide-PEG3-biotin conjugate has been used for the detection of a cellular alkyne cholesterol analog via click chemistry using fluorescence microscopy after incubation with a fluorescent streptavidin-conjugate. It has also been used to prepare a novel multifunctional benzophenone linker that can be used for pull-down assays, photoaffinity labeling and double-click stapling techniques.
Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it′s components do not react with the biological environment.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chapter Five-Mapping Lysine Acetyltransferase?Ligand Interactions by Activity-Based Capture
Montgomery DC & Meier JL
Methods in Enzymology, 574, 105-123 (2016)
A novel alkyne cholesterol to trace cellular cholesterol metabolism and localization.
Hofmann K, et al.
Journal of Lipid Research, 55(3), 583-591 (2014)
Development of a Multifunctional Benzophenone Linker for Peptide Stapling and Photoaffinity Labelling
Wu Y, et al.
Chembiochem, 17(8), 689-692 (2016)
Isotope-targeted glycoproteomics (IsoTaG) analysis of sialylated N-and O-glycopeptides on an Orbitrap Fusion Tribrid using azido and alkynyl sugars.
Woo CM, et al.
Analytical and Bioanalytical Chemistry, 409(2), 579-588 (2017)
Qingfei Zheng et al.
The Journal of organic chemistry, 85(3), 1691-1697 (2019-12-26)
Methylglyoxal (MGO) is a reactive dicarbonyl metabolite that modifies histones in vivo and induces changes in chromatin structure and function. Here we report the synthesis and application of a chemical probe for investigating MGO-glycation. A two-step synthesis of a Cu-click

Articles

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

See All

Related Content

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service