8.06020
Methylammonium chloride
for synthesis
Synonym(s):
Methylammonium chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CH6ClN
CAS Number:
Molecular Weight:
67.52
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-795-0
NACRES:
NA.22
Recommended Products
Quality Level
form
crystals
potency
1600 mg/kg LD50, oral (Rat)
pH
5-7 (20 °C, 10 g/L in H2O)
bp
225-230 °C/20 hPa
mp
228-233 °C
bulk density
1100 kg/m3
storage temp.
2-30°C
InChI
1S/CH5N.ClH/c1-2;/h2H2,1H3;1H
InChI key
NQMRYBIKMRVZLB-UHFFFAOYSA-N
Analysis Note
Assay (argentometric): ≥ 99.0 %
Melting range (lower value): ≥ 228 °C
Melting range (upper value): ≤ 233 °C
Identity (IR): passes test
Melting range (lower value): ≥ 228 °C
Melting range (upper value): ≤ 233 °C
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
408.7 °F - closed cup
Flash Point(C)
209.3 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service