Skip to Content
Merck
All Photos(1)

Key Documents

792640

Sigma-Aldrich

Potassium silicate, anhydrous

Synonym(s):

Potassium polysilicate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2O3Si
CAS Number:
Molecular Weight:
154.28
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

form

powder

mp

>300 °C

SMILES string

O=[Si](O[K])O[K]

InChI

1S/2K.O3Si/c;;1-4(2)3/q2*+1;-2

InChI key

NNHHDJVEYQHLHG-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mostafa M Rady et al.
Plant physiology and biochemistry : PPB, 139, 558-568 (2019-04-28)
The interplaying defensive roles of silicon (Si) and proline (Pro) in improving growth and yield attributes, physio-biochemical attributes, and antioxidant defense systems in common bean plant grown under saline (NaCl) and/or cadmium (Cd2+) stress were assessed. Seed were sown in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service