Skip to Content
Merck
All Photos(1)

Key Documents

742503

Sigma-Aldrich

Calcium silicate

purum, 12-22% Ca (as CaO) basis, ≥87% SiO2 basis

Synonym(s):

Calcium hydrosilicate, Calcium monosilicate, Calcium polysilicate, Calsilite, Silicic acid, calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22
form:
solid

grade

purum

Assay

≥87% SiO2 basis

form

solid

composition

Ca (as CaO), 12-22% T

loss

≤7.0% loss on drying, 2h; 105°C

particle size

7.0-10.0 μm

pH

9.5-11.5 (5% in H2O)

density

2.9 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-][Si]([O-])=O

InChI

1S/Ca.O3Si/c;1-4(2)3/q+2;-2

InChI key

OYACROKNLOSFPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Calcium silicate can be used as a precursor to synthesize several biomaterials. Examples for such applications include the preparation of biocompatible nanocomposites for bone tissue regeneration and the synthesis of sol-gel hybrid bioactive glasses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Salek et al.
Applied microbiology and biotechnology, 97(6), 2743-2752 (2012-11-29)
Silicate minerals can be a source of calcium and alkalinity, enabling CO2 sequestration in the form of carbonates. For this to occur, the mineral needs to be first dissolved in an acidifying process such as the biological process of anaerobic
Rania Hassan Fahmy
AAPS PharmSciTech, 13(3), 990-1004 (2012-07-19)
Multiparticulate floating drug delivery systems have proven potential as controlled-release gastroretentive drug delivery systems that avoid the "all or none" gastric emptying nature of single-unit floating dosage forms. An objective of the presence investigation was to develop calcium silicate (CaSi)/calcium
Emma C Goethals et al.
Langmuir : the ACS journal of surfaces and colloids, 29(2), 658-666 (2012-10-20)
Polymer nanocapsules have gained an important place as drug delivery vehicles for a myriad of biomedical applications. However, the influence of nanocapsule size, wall thickness, and porosity toward controlling the drug delivery efficiency of nanocapsular systems is not well understood.
Bobo Yu et al.
Langmuir : the ACS journal of surfaces and colloids, 28(50), 17465-17476 (2012-11-23)
The aim was to determine the most effective calcium precursor for synthesis of sol-gel hybrids and for improving homogeneity of sol-gel bioactive glasses. Sol-gel derived bioactive calcium silicate glasses are one of the most promising materials for bone regeneration. Inorganic/organic
Han Guo et al.
International journal of nanomedicine, 7, 3613-3624 (2012-08-01)
The purpose of this study was to synthesize a self-setting bioactive cement by incorporation of wollastonite nanofibers (WNFs) into calcium phosphate cement (CPC). The composition, morphology, setting time, compressive strength, hydrophilicity, and degradation of WNF-doped CPC (wnf-CPC) were investigated. Scanning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service