Skip to Content
Merck
All Photos(2)

Key Documents

S5904

Sigma-Aldrich

Sodium metasilicate nonahydrate

BioReagent, suitable for plant cell culture, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O3Si · 9H2O
CAS Number:
Molecular Weight:
284.20
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

Assay

≥98%

technique(s)

cell culture | plant: suitable

application(s)

agriculture

SMILES string

O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

InChI

1S/2Na.O3Si.9H2O/c;;1-4(2)3;;;;;;;;;/h;;;9*1H2/q2*+1;-2;;;;;;;;;

InChI key

PHIQPXBZDGYJOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silicon (Si) has been reported to effectively manage some pests and diseases of plants.Exogenous application of silicon (Si) in the form of sodium metasilicate reduced disease development caused by Penicillium expansum and Monilinia fructicola in sweet cherry fruit at 20 degrees C. The inhibition of fruit decay was correlated closely with Si concentrations.
Sodium metasilicate nonahydrate has been used as a trace element additive in bronchial epithelial growth medium (BEGM) and air-liquid interface (ALI) medium for culturing primary human airway epithelial (hAE) cells to study airway biology, disease, and therapy related to respiratory tract diseases.

Biochem/physiol Actions

Silicon (Si) has been reported to effectively manage some pests and diseases of plants. External application of silicon (Si) in the form of sodium metasilicate to Penicillium expansum and Monilinia fructicola infected sweet cherry fruit, at 20 °C, decreased the disease progression. The inhibition of fruit decay has been inferred by the Si concentrations.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pest risk assessment of Monilinia fructicola for the EU territory and identification and evaluation of risk management options
EFSA Journal, 9, 2119-2119 (2011)
Guo Z Qin et al.
Phytopathology, 95(1), 69-75 (2008-10-24)
ABSTRACT Exogenous application of silicon (Si) in the form of sodium metasilicate reduced disease development caused by Penicillium expansum and Monilinia fructicola in sweet cherry fruit at 20 degrees C. The inhibition of fruit decay was correlated closely with Si
Scott H Randell et al.
Methods in molecular biology (Clifton, N.J.), 742, 285-310 (2011-05-07)
When primary human airway epithelial (hAE) cells are grown in vitro on porous supports at an air-liquid interface (ALI), they recapitulate in vivo morphology and key physiologic processes. These cultures are useful for studying respiratory tract biology and diseases and
C Dugui-Es et al.
Communications in agricultural and applied biological sciences, 75(3), 497-505 (2010-01-01)
Silicon (Si) has been reported to effectively manage some pests and diseases of plants. This study was conducted to determine the effect of Si concentration, mode, and frequency of application in managing Meloidogyne incognita in cucumber. A susceptible cultivar of
M Leslie Fulcher et al.
Methods in molecular biology (Clifton, N.J.), 945, 109-121 (2012-10-26)
Human airway epithelial (hAE) cell cultures are instrumental for studying basic and applied aspects of respiratory tract biology, disease, and therapy. When primary epithelial cells from the human nasal passages or tracheo-bronchial airways are grown on porous supports at an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service