778346
Tungsten(IV) carbide
nanopowder, hexagonal, 150-200 nm, ≥99% trace metals basis
Synonym(s):
Tungsten monocarbide, Wolfram carbide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥99% trace metals basis
form
nanopowder
particle size
150-200 nm
SMILES string
[C-]#[W+]
InChI
1S/C.W/q-1;+1
InChI key
UONOETXJSWQNOL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Tungsten carbide is a well studied structural material with applications in coatings; composites and supports in heterogeneous catalysis. Tungsten carbide nanoparticles have been studied in wear resistant functional coatings as well as resilient composites.
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J. Catal., 134(2), 383-398 (1992)
Applied Physics Letters, 71(26), 3808-3810 (1997)
Advanced Engineering Materials, 2(5), 247-259 (2000)
J. Eur. Ceram. Soc., 25(1), 55-63 (2004)
Thin Solid Films, 370(1;2), 223-231 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service