Skip to Content
Merck
All Photos(2)

Key Documents

776742

Sigma-Aldrich

Silicon carbide

nanofiber, D <2.5 μm, L/D ≥ 20, 98% trace metals basis

Synonym(s):

Silicon carbide fibers, Silicon carbide nanowhiskers, Silicon carbide whiskers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiC
CAS Number:
Molecular Weight:
40.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98% trace metals basis

form

nanofiber

D

<2.5 μm

mp

2700 °C (lit.)

density

3.22 g/mL at 25 °C (lit.)

SMILES string

[C-]#[Si+]

InChI

1S/CSi/c1-2

InChI key

HBMJWWWQQXIZIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silicon carbide nanofibers are used with aluminum metal ceramic composites and ceramic composites to increase strength and reduce stress. Silicon carbide nanowhiskers can also be used in biocomposites for bone repair.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Strengthening of composites due to microstructural changes in the matrix
Arsenault, R. J.; Wang, L.; Feng, C. R.
Acta Metallurgica et Materialia, 39(1), 47-57 (1991)
Hockin H K Xu et al.
Biomaterials, 25(19), 4615-4626 (2004-05-04)
Self-hardening calcium phosphate cement (CPC) sets to form hydroxyapatite with high osteoconductivity, but its brittleness and low strength limit its use to only non-stress bearing locations. Previous studies developed bioactive composites containing hydroxyapatite fillers in Bis-GMA-based composites for bone repair
Fabrication of Mullite and Mullite-Matrix Composites by Transient Viscous Sintering of Composite Powders
Sacks, M.D.; Bozkurt, N.; Scheiffele, G.W.;
Journal of the American Ceramic Society. American Ceramic Society, 74(10), 2428-2437 (1991)

Articles

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service